To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:IgG1
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:0.5 mg
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below Avoid cycles of freezing and thawing. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:immunoaffinity chromatography
- Cat. no.:76226-084
Specifications
About this item
Camelids produce functional IgG isotypes that do not incorporate light chains. Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. At present, three subclasses of camelid IgG have been identified (IgG1, IgG2, IgG3), of which IgG2 and IgG3 lack the light chains. IgG1 binds strongly to protein A and G, is composed of conventional antibodies, and totals 25 % of serum IgG. Llama IgG1 antibodies migrate as a 150-kDa protein in SDS-polyacrylamide gel under non-reducing conditions. Under reducing conditions, IgG1 antibodies split into four proteins (two of each H- and L-chains), which gives protein bands of about 52-55 kDa (H-chain) and about 27 kDa (L-chain). Anti-Llama IgG1 generated in rabbit detects specifically the Llama IgG1 isotype. This anti-Llama secondary Antibody is suitable for western blot, ELISA, ChIP and immunohistochemistry as well as other more general immunoassays.
Antibody Anti-Llama IgG1 is suitable for immunoblotting (western or dot blot), ELISA, and immunohistochemistry assays requiring lot-to-lot consistency.
Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Camel