Order Entry
ContactUsLinkComponent
Anti-VhH Rabbit Polyclonal Antibody
Anti-VhH Rabbit Polyclonal Antibody
Catalog #: 76234-932
PAB RABBIT VHH IGG 25UL ELISA WEST BLOT
Anti-VhH Rabbit Polyclonal Antibody
Catalog #: 76234-932
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    VhH
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Protein A purified
  • Cat. no.:
    76234-932

Specifications

About this item

Rabbit Anti-VhH antibody can be used to detect single domain VhH antibodies. The single-domain antibody (sdAb) is a small (12 - 14 kDa) antibody fragment that consists of a monomeric variable domain derived from the heavy chain, also called a VhH antibody. These heavy chain only Fab-like domains have activity that is similar to a whole antibody, and they are able to bind to a specific antigens. SdAb's are derived from camelid species that include llamas, alpacas and camels. Camelids produce both classical (containing heavy and light chain fragments) and non-classical antibody structures (containing only a heavy chain). VhH antibodies are the smallest functional antigen-binding fragment that occurs in nature and these are now being used in biotechnology as a novel antibody scaffold. The small size of the VhH single domain antibody makes it very attractive for use in diagnostic imaging and they have potential for therapeutic activity.

Anti-VhH antibody has been tested by ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Some cross-reactivity to E.coli proteins may be observed.

Type: Primary
Antigen: VhH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought