Specifications
- Antibody type:Primary
- Antigen symbol:VhH
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western blot:Yes
- Size:100 µL
- Storage temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:multi-step process which includes delipidation and salt fractionation
- Cat. no.:76234-780
Specifications
About this item
Rabbit Anti-VhH antibody can be used to detect single domain VhH antibodies. The single-domain antibody (sdAb) is a small (12 - 14 kDa) antibody fragment that consists of a monomeric variable domain derived from the heavy chain, also called a VhH antibody. These heavy chain only Fab-like domains have activity that is similar to a whole antibody, and they are able to bind to a specific antigens. SdAb's are derived from camelid species that include llamas, alpacas and camels. Camelids produce both classical (containing heavy and light chain fragments) and non-classical antibody structures (containing only a heavy chain). VhH antibodies are the smallest functional antigen-binding fragment that occurs in nature and these are now being used in biotechnology as a novel antibody scaffold. The small size of the VhH single domain antibody makes it very attractive for use in diagnostic imaging and they have potential for therapeutic activity.
Anti-VhH antibody has been tested by ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Some cross-reactivity to E.coli proteins may be observed.
Type: Primary
Antigen: VhH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: