Order Entry
ContactUsLinkComponent
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Catalog # 76262-932
undefined
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Catalog # 76262-932
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TNFRSF10A
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Size:
    100 µg
  • Purification:
    Affinity purified
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Cat. no.:
    76262-932

Specifications

About this item

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms.  Apoptosis is induced by certain cytokines in the TNF family, including TNF and Fas ligand, through their death domain containing receptors, TNFR1 and Fas.  A novel death domain containing receptor was recently identified and designated DR4 (for death receptor 4).  The ligand for this novel death receptor has been identified and termed TRAIL, which is a new member in the TNF family. DR4 is also called TRAIL receptor-1 (TRAIL-R1). DR4 is expressed in most human tissues including spleen, peripheral blood leukocytes, small intestine and thymus. Like TNFR1, Fas and DR3, DR4 mediates apoptosis and NF-kB activation in many tissues and cells. Anti-DR4 antibody is ideal for investigators involved in Apoptosis and Cytokines and Growth Factors.

DR4 Antibody has been tested for use in ELISA, western blotting, and immunohistochemistry.  HeLa whole cell lysate can be used as a positive control for western blotting, and a band at approximately 57 kDa is expected. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TNFRSF10A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Frequently Bought Together