To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:TNFR-I
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:TNFR-I
- ELISA:Yes
- Size:100 µL
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:Protein G purified
- Cat. no.:76234-954
Specifications
About this item
Human TNF p55 Receptor Antibody recognizes TNFR1, the p55 soluble TNFR protein. sTNFRp55 is one of the two transmembrane receptors for tumor necrosis factor (TNF) with different molecular weight, 55 kD (p55) and 75 kD (p75), have been identified. The extracellular part of both receptors are shedded by proteolytic cleavage and exist as soluble receptors which bind to TNF in plasma and other biological fluids. sTNFR proteins have been implicated in certain leukemias. Anti-sTNFRp55 antibody is suitable for researchers in Cancer, Immunology, and Cell Biology.
Anti-TNF p55 Receptor (sTNFRp55) antibody has been tested for use in ELISA, radioimmunoassays, immunoblotting, flow cytometry and immunoprecipitation. Reactivity in other immunoassays is unknown. This product has been assayed for the ability to immunoprecipitate antigen and a dilution range of 1:400 to 1:800 is suggested. For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background. This product has been assayed by immunoblot and a working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against soluble (extracellular) sTNFRp55 at a dilution range of 1:2,000 to 1:8,000. For use in ELISA formats, this antibody is best used as the second antibody in combination with a monoclonal antibody as a capture antibody. This product has been assayed by radioimmunoassay against antigen. A dilution of 1:8,000 is suggested for this immunoassay.
Type: Primary
Antigen: TNFR-I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: