Order Entry
ContactUsLinkComponent
Anti-TNFR-I Rabbit Polyclonal Antibody
Anti-TNFR-I Rabbit Polyclonal Antibody
Catalog #: 76234-808
Anti-TNFR-I Rabbit Polyclonal Antibody
Catalog #: 76234-808
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    TNFR-I
  • ELISA:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 µL
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Purification:
    heated to 56°C
  • Cat. no.:
    76234-808

Specifications

About this item

Tumor necrosis factor receptor superfamily member 1A (TNF p55 Receptor) is a receptor for TNFSF2/TNF-alpha and homotrimeric TNFSF1/lymphotoxin-alpha. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. It contributes to the induction of non-cytocidal TNF effects including anti-viral state and activation of the acid sphingomyelinase.

This antiserum against Human TNF p55 Receptor (sTNFRp55) has been tested for use in ELISA,  radioimmunoassays, immunoblotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.This product has been assayed for the ability to immunoprecipitate antigen.  A dilution range of 1:400 to 1:800 is suggested for this immunoassay.  For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background.  This product has been assayed by immunoblot  using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:200 to 1:400 is suggested for this application.  This product has been assayed by ELISA against soluble (extracellular) sTNFRp55 using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution range of 1:1,000 to 1:2,000 is suggested for this product.  For use in ELISA formats, this antibody is best used as the second antibody  in combination with a monoclonal antibody as a capture antibody.  This product has been assayed by radioimmunoassay against antigen.  A dilution of 1:8,000 is suggested for this immunoassay.

Type: Primary
Antigen: TNFR-I
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought