Order Entry
ContactUsLinkComponent
Anti-TNFA Rabbit Polyclonal Antibody
Anti-TNFA Rabbit Polyclonal Antibody
Catalog #: 76234-806
PAB RAB TNF ALPHA HUMAN ANTISERUM 25UL
Anti-TNFA Rabbit Polyclonal Antibody
Catalog #: 76234-806
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    TNFA
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    heated to 56°C
  • Cat. no.:
    76234-806

Specifications

About this item

Anti TNF alpha Antibody recognizes TNF alpha (TNF, cachexin, cachectin, tumor necrosis factor-alpha or TNF-α) a cytokine involved in systemic inflammation. TNF alpha is a member of a group of cytokines that stimulate the acute phase reaction. It is produced chiefly by activated macrophages, although it can be produced by other cell types as well. The primary role of TNF alpha is in the regulation of immune cells. TNFis an endogenous pyrogen that is able to induce fever, apoptotic cell death, sepsis (through IL-1 & IL-6 production), cachexia, inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNF production has been implicated in a variety of human diseases, including Alzheimer's disease, cancer, major depression, and inflammatory bowel disease (IBD).

Anti-Human TNFα has been tested for use in ELISA, immunohistochemistry and immunoblotting. It recognizes the 17,000 MW TNFα. Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought