Order Entry
ContactUsLinkComponent
Anti-SLC7A11 Rabbit Polyclonal Antibody
Anti-SLC7A11 Rabbit Polyclonal Antibody
Catalog #: 76262-708
PAB RABBIT XCT HUMAN IGG 25UL WEST BLOT
Anti-SLC7A11 Rabbit Polyclonal Antibody
Catalog #: 76262-708
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    SLC7A11
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-708

Specifications

About this item

xCT belongs to the amino acid-polyamine-organocation (APC) superfamily and L-type amino acid transporter (LAT) family. xCT gene encodes a member of a heteromeric, sodium-independent, anionic amino acid transport system that is highly specific for cysteine and glutamate. In this system, designated Xc(-), the anionic form of cysteine is transported in exchange for glutamate. Increased expression of this gene results in neuronal cell death. xCT may be associated with kaposi sarcoma, dyscalculia, cystinuria, spondylolsis, and anemia of prematurity. Anti-xCT Antibody is useful for researchers interested in Cancer Research, Glucose/Energy Metabolism Research, and Cell Surface Protein Interaction Research.

Anti-xCT Antibody has been tested in Western Blot, ELISA, Immunofluorescence, and flow cytometry. Expect a band at 55.4 kDa in western blot using appropriate lysates. Positive control used HeLa, NIH-3T3, A549, HCT-116, or PC3 whole cell lysates at 0.1-1.0µg/mL for WB and PC3 with MeOH at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry and immunoprecipitation.

Type: Primary
Antigen: SLC7A11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought