Order Entry
ContactUsLinkComponent
Anti-PECAM1 Rabbit Polyclonal Antibody
Anti-PECAM1 Rabbit Polyclonal Antibody
Catalog #: 76234-788
PAB RAB CD31 RAT MOUSE HUMAN IGG 100UL
Anti-PECAM1 Rabbit Polyclonal Antibody
Catalog #: 76234-788
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    PECAM1
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 µL
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Purification:
    multi-step process which includes delipidation and salt fractionation
  • Cat. no.:
    76234-788

Specifications

About this item

Anti-CD31 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). CD31 (PECAM-1) is a 130 kDa platelet endothelial cell adhesion molecule encoded by the PECAM1 gene found on chromosome 17 in humans. CD31 is expressed on platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions. CD31 plays a key role in modulation of integrin-mediated cell adhesion, leukocyte transendothelial migration, angiogenesis, apoptosis and macrophage phagocytosis. CD31 is also expressed in certain tumors, including epithelioid hemangioendothelioma, epithelioid sarcoma-like hemangioendothelioma, other vascular tumors, histiocytic malignancies, and plasmacytomas. Anti-CD31 is ideal for researchers interested in Stem Cell Research, Epigenetics, and Cell-cycle Regulation research.

CD31 antibody has been tested for use in ELISA, immunohistochemistry, and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80 kDa in size corresponding to CD31 protein by western blotting in the appropriate stimulated tissue or cell lysate or extract.

Type: Primary
Antigen: PECAM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought