Order Entry
ContactUsLinkComponent
Anti-Pax7 Rabbit Polyclonal Antibody
Anti-Pax7 Rabbit Polyclonal Antibody
Catalog #: 76262-702
PAB RABBIT PAX7 MOUSE IGG 100 UG ELISA
Anti-Pax7 Rabbit Polyclonal Antibody
Catalog #: 76262-702
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    Pax7
  • ELISA:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-702

Specifications

About this item

The Anti-Pax7 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Paired box protein Pax-7 transcription factor plays a role in myogenesis through regulation of muscle precursor cells proliferation. These genes play critical roles during fetal development and cancer growth. It is expressed specifically during the development of the nervous and muscular system. Anti-Pax7 antibody is ideal for researchers interested in developmental biology, stem cell research, cell growth and cancer research.

Anti-Pax7 antibody has been tested in ELISA and Western Blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 54.9 kDa in size corresponding to Pax7 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: Pax7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought