Order Entry
ContactUsLinkComponent
Anti-PAX6 Mouse Monoclonal Antibody [Clone: 7H10.D3.D4.G5]
Anti-PAX6 Mouse Monoclonal Antibody [Clone: 7H10.D3.D4.G5]
Catalog #: 76235-002
MAB MS PAX6 7H10.D3.D4.G5 HU IGG1 100UG
Anti-PAX6 Mouse Monoclonal Antibody [Clone: 7H10.D3.D4.G5]
Catalog #: 76235-002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Clone:
    7H10.D3.D4.G5
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Antigen symbol:
    PAX6
  • ELISA:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Purification:
    Protein A purified
  • Cat. no.:
    76235-002

Specifications

About this item

Paired box protein Pax-6 also known as aniridia type II protein (AN2) or oculorhombin is a transcription factor with key functions in the development of the eye, nose, central nervous system and pancreas. Pax6 further seems to be required for the differentiation of pancreatic islet alpha cells. PAX6 has been shown to bind to a promoter element in the glucagon, insulin and somatostatin promoters and regulates the specification of the ventral neuron subtypes by establishing the correct progenitor domains. Isoform 5a appears to function as a molecular switch that specifies target genes.
Anti-PAX6 antibody is ideal for researchers interested in developmental biology, stem cell research, cell growth and cancer research.

Anti-PAX6 antibody is suitable for ELISA and Immunofluorescence.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 46.7 kDa in size corresponding to PAX6 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PAX6
Clonality: Monoclonal
Clone: 7H10.D3.D4.G5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Customers Who Bought This Also Bought