To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:17C9.E1.H2.D7.H7.D5
- Host:Mouse
- Isotype:IgG1 lambda
- Antigen symbol:MAP2K1/MAP2K2
- ELISA:Yes
- Size:25 µL
- Western blot:Yes
- Modification:Phosphorylation
- Shipping temperature:Dry Ice
- Purification:Protein A purified
- Cat. no.:76235-000
Specifications
About this item
MEK1 pS222/MEK2 pS226 antibody detects MEK1 and MEK2. Mitogen-activated protein kinase kinase 1, (also known as MKK or MEK1), and Mitogen-activated protein kinase kinase 2, (also known as MEK2 or MKK2), are integral components of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 1 and 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. The MEK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
Anti-MEK1 pS222/MEK2 pS226 (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 43.5 kDa.
Type: Primary
Antigen: MAP2K1/MAP2K2
Clonality: Monoclonal
Clone: 17C9.E1.H2.D7.H7.D5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: