Order Entry
ContactUsLinkComponent
Anti-MAP2K1 Mouse Monoclonal Antibody (DL800) [Clone: 13B6.G12]
Anti-MAP2K1 Mouse Monoclonal Antibody (DL800) [Clone: 13B6.G12]
Catalog #: 76235-060
MAB MEK1 C-TERM 13B6.G12 HU DY800 100UG
Anti-MAP2K1 Mouse Monoclonal Antibody (DL800) [Clone: 13B6.G12]
Catalog #: 76235-060
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Clone:
    13B6.G12
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Antigen symbol:
    MAP2K1
  • Conjugation:
    DyLight® 800
  • ELISA:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    Protein A purified
  • Cat. no.:
    76235-060

Specifications

About this item

MEK1 antibodies detect the MEK1 isoform.  Mitogen-activated protein kinase kinase 1, also known as MKK or MEK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 1 DyLight™800 Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA.  Specific conditions of reactivity should be optimized by the end user.  Expect a band of approximately 43.5 kDa.

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 13B6.G12
Conjugation: DL800
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Customers Who Bought This Also Bought