Order Entry
ContactUsLinkComponent
Anti-Leptin Rabbit Polyclonal Antibody
Anti-Leptin Rabbit Polyclonal Antibody
Catalog #: 76234-822
Anti-Leptin Rabbit Polyclonal Antibody
Catalog #: 76234-822
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    Leptin
  • ELISA:
    Yes
  • Size:
    100 µL
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Purification:
    heated to 56°C
  • Cat. no.:
    76234-822

Specifications

About this item

Leptin is a key player in the regulation of energy balance and body weight control. Once released into the circulation, it has central and peripheral effects by binding LEPR, found in many tissues, which results in the activation of several major signaling pathways. It acts as an appetite-regulating factor, regulates bone mass and secretion of hypothalamo-pituitary-adrenal hormones, it increases basal metabolism, influences reproductive function, regulates pancreatic beta-cell function and insulin secretion, it induces FOS and SOCS3 expression to release anorexigenic peptides. It has a modulatory role in nutrient absorption. It reduces glucose absorption by enterocytes by activating PKC and leading to a sequential activation of p38, PI3K and ERK signaling pathways which exerts an inhibitory effect on glucose absorption. It acts as a growth factor on certain tissues, through the activation of different signaling pathways increases expression of genes involved in cell cycle regulation such as CCND1, via JAK2-STAT3 pathway, or VEGFA, via MAPK1/3 and PI3K-AKT1 pathways. It may also play an apoptotic role via JAK2-STAT3 pathway and up-regulation of BIRC5 expression. It plays a pro-inflammatory role, in synergy with IL1B, by inducing NOS2 which promotes the production of IL6, IL8 and Prostaglandin E2, through a signaling pathway that involves JAK2, PI3K, MAP2K1/MEK1 and MAPK14/p38. In adaptive immunity, promotes the switch of memory T-cells towards T helper-1 cell immune responses. Increases CD4+CD25- T cells proliferation and reduces autophagy during TCR (T cell receptor) stimulation, through MTOR signaling pathway activation and BCL2 up-regulation.

This antiserum against Mouse Leptin has been tested for use in ELISA and immunoblotting.  Reactivity in other immunoassays is unknown.  Obesity associated leptin is the product of the OB gene and has been identified with Type II diabetes.This product has been assayed by immunoblot against tissue homogenates using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate.  A working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against recombinant mouse leptin using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature.  A working dilution of 1:2,400 is suggested for this product. 

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought