To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:IL1RA
- ELISA:Yes
- Size:100 µL
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:DEAE fractionation
- Cat. no.:76235-122
Specifications
About this item
Interleukin 1 receptor antagonist is secreted by various types of cells including immune, epithelial and adipocytes. IL 1Ra is used to treat rheumatoid arthritis. Anti-IL-1 RA antibody is ideal for investigators involved in Cancer and Immunology research.
This IgG fraction antibody of anti-Human IL-1 Receptor Antagonist has been tested for use in neutralizations, ELISA, radioimmunoassays, western blotting, immunohistochemistry and flow cytometry. Reactivity in other immunoassays is unknown.This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate. A working dilution range of 1:400 to 1:800 is suggested for this application. This product has been assayed by ELISA against IL-1Receptor Antagonist using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature. A working dilution range of 1:500 to 1:1,000 is suggested for this product. For use in ELISA formats, this antibody is best used as the second antibody in combination with a monoclonal antibody as a capture antibody. This product has been assayed by immunohistochemistry. A dilution of 1:400 or greater is suggested for this immunoassay. Either paraffin fixation or cryofixation can be used for immunohistochemistry. This product has been assayed by radioimmunoassay against antigen. A dilution of 1:8,000 is suggested for this immunoassay.
Type: Primary
Antigen: IL1RA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: