Order Entry
ContactUsLinkComponent
Anti-IL1A Rabbit Polyclonal Antibody
Anti-IL1A Rabbit Polyclonal Antibody
Catalog # 76235-118
undefined
Anti-IL1A Rabbit Polyclonal Antibody
Catalog # 76235-118
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IL1A
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Size:
    100 µL
  • Purification:
    DEAE fractionation
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Cat. no.:
    76235-118

Specifications

About this item

Interleukin 1 alpha is produced by activated macrophages. Anti-IL-1 alpha antibody is ideal for investigators involved in Immunology research.

This IgG fraction antibody of anti-Human IL-1a has been tested for use in neutralizations, ELISA,  immunohistochemistry, flow cytometry and immunoblotting.  It recognizes the 17,000 MW mature IL-1a.  Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Frequently Bought Together