Order Entry
ContactUsLinkComponent
Anti-IKBKG Rabbit Polyclonal Antibody
Anti-IKBKG Rabbit Polyclonal Antibody
Catalog #: 76234-934
PAB RAB NEMO/IKK-GAMMA HUMAN IGG 100UG
Anti-IKBKG Rabbit Polyclonal Antibody
Catalog #: 76234-934
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    IKBKG
  • ELISA:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
  • Cat. no.:
    76234-934

Specifications

About this item

Anti-NEMO antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Anti-NEMO antibody detects recombinant and endogenous NEMO. NEMO, the regulatory subunit of the IKK core complex, phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. Its binding to scaffolding polyubiquitin seems to play a role in IKK activation by multiple signaling receptor pathways. Nemo is also considered to be a mediator for TAX activation of NF-kappa-B and may be implicated in NF-kappa-B-mediated protection from cytokine toxicity. NEMO is essential for viral activation of IRF3 and involved in TLR3- and IFIH1-mediated antiviral innate response. The innate antivaral response from NEMO requires 'Lys-27'-linked polyubiquitination. Anti-NEMO is ideal for researchers interested in Immunology and Cancer research.

Anti-NEMO antibody is suitable for western blotting, immunoprecipitation, and ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 48kDa in size corresponding to endogenous NEMO protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: IKBKG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought