Order Entry
ContactUsLinkComponent
Anti-HIST1H3A Rabbit Polyclonal Antibody
Anti-HIST1H3A Rabbit Polyclonal Antibody
Catalog #: 76234-928
PAB RABBIT HISTONE H3 HUMAN IGG 25UL WB
Anti-HIST1H3A Rabbit Polyclonal Antibody
Catalog #: 76234-928
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    HIST1H3A
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Protein A purified
  • Cat. no.:
    76234-928

Specifications

About this item

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Histone proteins are highly post-translationally modified with Histone H3 being the most extensively modified of the five histones. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H3 Antibody is ideal for investigators involved in Cell Signaling, Epigenetics, Nuclear Signaling research and Signal Transduction research.

Histone H3 antibody has been tested for use in ELISA, IHC, and western blot. For western blots expect a band of approximately 15.4 kDa in size corresponding tothe Histone 3 protein. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought