Order Entry
ContactUsLinkComponent
Anti-HbD Mouse Monoclonal Antibody [Clone: 21G1.F1.B9.G9.D11]
Anti-HbD Mouse Monoclonal Antibody [Clone: 21G1.F1.B9.G9.D11]
Catalog #: 76234-996
MAB HBB A-2 21G1.F1.B9.G9.D11 HUMAN 25UL
Anti-HbD Mouse Monoclonal Antibody [Clone: 21G1.F1.B9.G9.D11]
Catalog #: 76234-996
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Clone:
    21G1.F1.B9.G9.D11
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Antigen symbol:
    HbD
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Protein A purified
  • Cat. no.:
    76234-996

Specifications

About this item

HbA-2 or hemoglobin delta subunit antibodies detect the delta-specific sequence in the hemoglobin delta-subunit found in HbA-2. Functional hemoglobin (Hb) is a hetero tetramer and the dominant form of Adult Hb is composed of 2 alpha and 2 beta subunits (α2β2). Hemoglobin A-2 (HbA-2) is a normal but minor variant of hemoglobin A that consists of two alpha and two delta chains (α2δ2). Hemoglobin A-2 may be increased in beta thalassemia or in people who are heterozygous for the beta thalassemia gene, and HbA2 is also linked to neurological disorders. HbA-2 form exists in small amounts in all adult humans (1.5-3.1% of all hemoglobin molecules) and is increased in people with Sickle-cell disease. Its normal biological role is not well understood. HbA-2 antibody does not react other forms of Hb including no cross-reaction to HbA or beta subunit. This antibody is ideal for investigators involved in Cardiovascular and developmental biology research.

Anti-Hemoglobin beta A-2 (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 16 kDa.

Type: Primary
Antigen: HbD
Clonality: Monoclonal
Clone: 21G1.F1.B9.G9.D11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Customers Who Bought This Also Bought