To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:20C11.H5.E11
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen symbol:GSTM3
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:100 µg
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:Protein A purified
- Cat. no.:76234-894
Specifications
About this item
Rockland produces a wide range of human GST antibodies in our laboratories. Select appropriate GST antibodies for your research by isotype, epitope, applications and species reactivity. There are 22 members of the human GST family of proteins. GST is responsible for the conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. The amino acid sequence GST is highly conserved in most organisms including mammals. GSTs proteins are typically homodimeric, with both heterologous GST dimers have been observed. GST monomers have an average molecular weight of approximately 25-28 kDa in size. Note a different form of non-human GST (Glutathione-S-Transferase) is used as a protein expression tag commonly in molecular biology applications. All anti-GST antibodies my not react with recombinant GST-fusion proteins.
This antibody is useful in ELISA and Wetern Blot. Suitable for most immunological techniques requiring high titer binding and lot-to-lot consistency. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: GSTM3
Clonality: Monoclonal
Clone: 20C11.H5.E11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: