Order Entry
ContactUsLinkComponent
Anti-F11R Rabbit Polyclonal Antibody
Anti-F11R Rabbit Polyclonal Antibody
Catalog #: 76262-670
PAB RABBIT JAM A PY280 HUMAN IGG 25UL WB
Anti-F11R Rabbit Polyclonal Antibody
Catalog #: 76262-670
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    F11R
  • ELISA:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Modification:
    Phosphorylation
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-670

Specifications

About this item

Anti-JAM A pY280 was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). JAM A pY280 Antibody detects junctional adhesion molecule A (JAM A) protein phosphorylated at the tyrosine 280 location. JAM A is involved in cell-cell contact and tight junction formation in epithelial cells. JAM A protein is a transmembrane protein expressed in epithelial and endothelial cells as well as in hematopoietic cells and is important for a variety of cellular processes including tight junction assembly, leukocyte transmigration, platelet activation, angiogenesis and virus binding. In addition, JAM A may be a potential therapeutic target for breast cancer.

This affinity purified antibody has been tested for use in ELISA, IF, and western blot.   Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 32.5 kDa in size corresponding to JAM A by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: F11R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought