Order Entry
ContactUsLinkComponent
Anti-COL6A1 Rabbit Polyclonal Antibody (HRP)
Anti-COL6A1 Rabbit Polyclonal Antibody (HRP)
Catalog #: 76262-812
PAB RAB COLLAGEN TYPE VI MAMMAL HRP 50UG
Anti-COL6A1 Rabbit Polyclonal Antibody (HRP)
Catalog #: 76262-812
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    COL6A1
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    immunoaffinity chromatography
  • Cat. no.:
    76262-812

Specifications

About this item

Collagen Type VI Peroxidase Conjugated Antibody is specific for collagen type VI protein. The collagens are a superfamily of proteins that play a role in maintaining the integrity of various tissues. Collagen VI is a major structural component of microfibrils. The basic structural unit of collagen VI is a heterotrimer of the alpha1(VI), alpha2(VI), and alpha3(VI) chains. The alpha2(VI) and alpha3(VI) chains are encoded by the COL6A2 and COL6A3 genes, respectively. The protein encoded by this gene is the alpha 1 subunit of type VI collagen (alpha1(VI) chain). Mutations in the genes that code for the collagen VI subunits result in the autosomal dominant disorder, Bethlem myopathy.

Anti-Collagen Type VI (RABBIT) Peroxidase Conjugated Antibody is suitable for western blot, immunoprecipitatioin, and immunohistochemistry. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL6A1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought