Order Entry
ContactUsLinkComponent
Anti-COL4A4 Rabbit Polyclonal Antibody (FITC)
Anti-COL4A4 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-772
Anti-COL4A4 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-772
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    COL4A4
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    immunoaffinity chromatography
  • Cat. no.:
    76262-772

Specifications

About this item

Anti-Collagen Type IV Fluorescein Conjugated Antibody detects collagen. Type-IV collagen is a type of collagen found primarily in the basal lamina. It is the major structural element of basal membranes. Anti-Collagen Type IV antibody is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.

Anti-Collagen Type IV Fluorescein Conjugated Antibody is suitable for western blotting, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL4A4
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought