Order Entry
ContactUsLinkComponent
Anti-COL2A1 Rabbit Polyclonal Antibody (FITC)
Anti-COL2A1 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-768
PAB COLLAGEN TYPE II MAMMAL FITC 50UG WB
Anti-COL2A1 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-768
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    COL2A1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    immunoaffinity chromatography
  • Cat. no.:
    76262-768

Specifications

About this item

Anti-Collagen Type II Fluorescein Conjugated Antibody detects Type-II collagen which is the basis for articular cartilage and hyaline cartilage. It makes up 50% of all protein in cartilage and 85-90% of collagen of articular cartilage. Type II collagen does form fibrils which allow cartilage to entrap the proteoglycan aggregate as well as provide tensile strength to the tissue. Anti-Collagen Type II antibody is ideal for investigators involved in Immunology, Neuroscience and Signal Transduction research.

Anti-Collagen Type II Fluorescein Conjugated Antibody is suitable for western blotting, IP, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL2A1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought