Order Entry
ContactUsLinkComponent
Anti-COL1A1 Rabbit Polyclonal Antibody (HRP)
Anti-COL1A1 Rabbit Polyclonal Antibody (HRP)
Catalog #: 76262-802
PAB RAB COLLAGEN TYPE I MAMMAL HRP 50UG
Anti-COL1A1 Rabbit Polyclonal Antibody (HRP)
Catalog #: 76262-802
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    COL1A1
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    immunoaffinity chromatography
  • Cat. no.:
    76262-802

Specifications

About this item

COLLAGEN Type I Peroxidase Conjugated Antibody is specific for Collagen Type I. Collagen Type I (Type-I collagen) is the most abundant collagen of the human body. It is present in scar tissue, the end product when tissue heals by repair. It is also found in tendons, the endomysium of myofibrils and the organic part of bone. Anti-collagen Type I antibody is suitable for Cancer research and other general research.

Anti-COLLAGEN Type I Peroxidase Conjugated Antibody is suitable for western blot, immunoprecipitatioin, Flow Cytometry, and immunohistochemistry. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought