Order Entry
ContactUsLinkComponent
Anti-CD44 Rabbit Polyclonal Antibody
Anti-CD44 Rabbit Polyclonal Antibody
Catalog #: 76263-056
Anti-CD44 Rabbit Polyclonal Antibody
Catalog #: 76263-056
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    CD44
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76263-056

Specifications

About this item

CD44 was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). CD44 is a receptor for hyaluronic acid (HA), an integral component of the extracellular matrix. CD44 mediates cell-cell and cell-matrix interactions through its affinity for HA, and can also interact with ligands such as osteopontin, collagens, and matrix metalloproteinases (MMPs). The multiple protein isoforms are encoded by a single gene by alternative splicing and are further modified by a range of post-translational modifications. CD44 function is controlled by these posttranslational modifications. The major physiological role of CD44 is to maintain organ and tissue structure via cell-cell and cell-matrix adhesion, but certain variant isoforms can also mediate lymphocyte activation and homing, and the presentation of chemical factors and hormones. CD44 participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. CD44 is a multi-structural and multi-functional cell surface molecule involved in cell proliferation, cell differentiation, cell migration, angiogenesis, presentation of cytokines, chemokines, and growth factors to the corresponding receptors, and docking of proteases at the cell membrane, as well as in signaling for cell survival. All these biological properties are essential to the physiological activities of normal cells, but they are also associated with the pathologic activities of cancer cells. CD44, particularly its variants, may be useful as a diagnostic or prognostic marker of malignancy and, in at least some human cancers, it may be a potential target for cancer therapy.

Anti-CD44 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 81.5 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought