Order Entry
ContactUsLinkComponent
Anti-CCL3 Rabbit Polyclonal Antibody
Anti-CCL3 Rabbit Polyclonal Antibody
Catalog #: 76234-818
Anti-CCL3 Rabbit Polyclonal Antibody
Catalog #: 76234-818
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    CCL3
  • ELISA:
    Yes
  • Size:
    100 µL
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Purification:
    heated to 56°C
  • Cat. no.:
    76234-818

Specifications

About this item

C-C motif chemokine 3, CCL3 or (MIP-1a) is a monokine with inflammatory and chemokinetic properties. CCL3 binds to CCR1, CCR4 and CCR5. It is one of the major HIV-suppressive factors produced by CD8+ T-cells. Recombinant MIP-1-alpha induces a dose-dependent inhibition of different strains of HIV-1, HIV-2, and simian immunodeficiency virus (SIV).

This antiserum against Human MIP-1a has been tested for use in ELISA of human MIP-1a found in cell supernatants or in plasma or serum.  Reactivity in other immunoassays is unknown.

Type: Primary
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought