Order Entry
ContactUsLinkComponent
Anti-BCL3 Rabbit Polyclonal Antibody
Anti-BCL3 Rabbit Polyclonal Antibody
Catalog #: 76262-712
PAB RABBIT BCL3 HUMAN IGG 25 UL ELISA WB
Anti-BCL3 Rabbit Polyclonal Antibody
Catalog #: 76262-712
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    BCL3
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-712

Specifications

About this item

BCL3 belongs to the ANKRD family. The protein contains seven ankyrin repeats, which are closely related to those found in I kappa B proteins. BCL3 functions as a transcriptional co-activator, regulator of transcriptional activation, promotor of transcription, and inhibitor of nuclear translocation of NF-kappa B. It is a proto-oncogene identified by its translocation into the immunoglobulin alpha-locus in some cases of B-cell leukemia. BCL3 contributes to the regulation of cell proliferation. Therefore, BCL3 may be associated with B-cell chronic lymphocytic leukemia, lymphoma, or other hematologic diseases. Anti-BCL3 Antibody is useful for researchers interested in Cancer Research, NF-Kappa B Pathway, and TNF Signaling Pathway.

Anti-BCL-3 Antibody has been tested in Western Blot, ELISA, and Immunofluorescence. Expect a band at 47.5 kDa in western blot using appropriate lysates. Positive control used recombinant BCL3 protein, HEK293T overexpressing BCL3 or U87 whole cell lysates at 1µg/mL in WB; and U87MG with PFA at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry, flow cytometry, and immunoprecipitation.

Type: Primary
Antigen: BCL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought