Order Entry
ContactUsLinkComponent
Anti-AKT1 Rabbit Polyclonal Antibody
Anti-AKT1 Rabbit Polyclonal Antibody
Catalog #: 76262-930
PAB RAB AKT PHOSPHO T308 HUMAN IGG 25UL
Anti-AKT1 Rabbit Polyclonal Antibody
Catalog #: 76262-930
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    AKT1
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Modification:
    Phosphorylation
  • Shipping temperature:
    Dry Ice
  • Purification:
    immunoaffinity chromatography
  • Cat. no.:
    76262-930

Specifications

About this item

AKT is a component of the PI-3 kinase pathway and is activated by phosphorylation at  Ser 473 and Thr 308.  AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and rac (related to A and C kinases). AKT is a key regulator of many signal transduction pathways.  AKT Exhibits tight control over cell proliferation and cell viability.  Overexpression or inappropriate activation of AKT is noted in many types of cancer.   AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus.  AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation.

AKT antibody is phospho specific for pT308 and is suitable for western blotting, Flow Cytometry, and ELISA.

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought