SRT®-C SEC HPLC Column, Sepax Technologies
Catalog # 103061-088
Supplier: Sepax Technologies
Specifications
- Column Format:Analytical
- Ø int.:7.80 mm
- Particle Size:5 µm
- Pore size:100 Å
- Max. pressure:~ 4500 psi
- Length:250 mm
- pH range:2 - 8.5
- Temperature limit:~ 80 °C
- Cat. no.:103061-088
- Packing Material:Neutral, hydrophilic film bonded silica
- Phase:SRT®-C SEC
- Supplier No.:235100-7825
Specifications
About this item
SRT®-C SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica.
- High separation efficiency and resolution
- High capacity
- High lot-to-lot reproducibility
- High protein recovery with intact biological activity
- Negligible non-specific interactions
SRT®-C, uses lay-down monolayer on porous silica as oppose to SRT of stand-up monolayer, offers ideal phase chemistry for hydrophobic samples like insulin, membrane protein monoclonal antibodies derivatized with polymer branches (polypeptide or PEG) separation.