Ultrabake™ Silicone Caps, SC-SH Series, Caplugs
Supplier: Caplugs
Certificates
About this item
SC-SH Series Ultrabake™ Silicone Caps are designed to mask plain and threaded studs, tube ends and even slotted holes; they will mask off the stud and allow finishing up to it. Recommended for powder coating, e-coating, plating and anodizing.
- Recommended temperatures up to 600 °F (315 °C)
- Rugged and durable
- Can stretch to mask larger dimensions
- Reusable
Frequently Bought Together