Order Entry
Canada
ContactUsLinkComponent
SSPE Buffer, 20X, Rockland Immunochemicals
SSPE Buffer, 20X, Rockland Immunochemicals
Catalog # 76236-412
CAS Number:  
SSPE Buffer, 20X, Rockland Immunochemicals
Catalog # 76236-412
Supplier Number:  MB-044-4000
CAS Number:  

Specifications

  • Description:
    20X SSPE Buffer
  • Size:
    4 L
  • Cat. no.:
    76236-412

Specifications

About this item

Saline–Sodium Phosphate–EDTA (SSPE) is used in bacterial screening and hybridization procedures.

  • 20X concentrated stock solution - DEPC Treated
  • 3.0 M Sodium Chloride, 0.2 M Sodium Hydrogen Phosphate, 0.02 M EDTA at a pH of 7.4
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers

The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Application Note: This product is a 20X concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 3.0 M Sodium Chloride, 0.2 M Sodium Hydrogen Phosphate, 0.02 M EDTA at a pH of 7.4.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water treated with diethyl pyrocarbonate (DEPC).