Order Entry
Canada
ContactUsLinkComponent
Mouse VEGF-B ELISA Kit, Rockland Immunochemicals
Mouse VEGF-B ELISA Kit, Rockland Immunochemicals
  76236-236
 :  
Mouse VEGF-B ELISA Kit, Rockland Immunochemicals
  76236-236
 :  KOA0852
 :  

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    VEGF-B
  • Description:
    Mouse VEGF-B ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-236

 

 

Mouse VEGF-B AccuSignal ELISA Kit

  • Natural and recombinant mouse VEGF-B.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: Q20-A207&Q20-R148

Vascular endothelial growth factor B also known as VEGF-B is a protein that, in humans, is encoded by the VEGF-B gene. VEGF-B is a growth factor that belongs to the vascular endothelial growth factor family, of which VEGF-A is the best-known member. In contrast to VEGF-A, VEGF-B plays a less pronounced role in the vascular system: Whereas VEGF-A is important for the formation of blood vessels, such as during development or in pathological conditions, VEGF-B seems to play a role only in the maintenance of newly formed blood vessels during pathological conditions. It also plays an important role on several types of neurons. And it is important for the protection of neurons in the retina and thecerebral cortex during stroke and of motoneurons during motor neuron diseases such as amyotrophic lateral sclerosis.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse VEGF-B standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin) to each empty well. We recommend that each mouse VEGF-B standard solution and each sample is measured in duplicate.