Specifications
- Primary antibody reactivity:Human
- Description:Human CD97 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-640
Specifications
About this item
Human CD97 AccuSignal ELISA Kit
- Natural and recombinant human CD97.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q21-Q398
Cluster of differentiation 97 is a protein also known as BL-Ac [F2] encoded by the ADGRE5 gene. The CD97 gene is mapped to 19p13.2-p13.12 by fluorescence in situ hybridization. In the immune system, CD97 is known as a critical mediator of host defense. Upon lymphoid, myeloid cells and neutrophil activation, CD97 is upregulated to promote adhesion and migration to sites of inflammation. Moreover, it has been shown that CD97 regulates granulocyte homeostasis. CD97 is expressed at the sarcoplasmic reticulum and the peripheral sarcolemma in skeletal muscle. However, lack of CD97 only affects the structure of the sarcoplasmic reticulum, but not the function of skeletal muscle.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml human CD97 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CD97 standard solution and each sample be measured in duplicate.