- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Target protein:IL-20
- Description:Mouse IL-20 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-504
Mouse Interleukin-20 AccuSignal ELISA Kit
- Natural and recombinant mouse IL-20.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: L25-E176
IL-20, also known as Interleukin 20 or Zcyto10, is a protein that in humans is encoded by the IL20 gene. This gene is mapped to 1q32.1. IL-20 is a protein belonging to the IL-10 family of cytokines. This cytokine has been shown to transduce its signal through signal transducer and activator of transcription 3 (STAT3) in keratinocytes. A specific receptor for this cytokine is found to be expressed in skin and upregulated dramatically in psoriatic skin, suggesting a role for this protein in epidermal function and psoriasis. IL-20 regulates proliferation and differentiation of keratinocytes during inflammation, particularly inflammation associated with the skin. In addition, IL-20 also causes cell expansion of multipotential hematopoietic progenitor cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse IL-20 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-20 standard solution and each sample be measured in duplicate.