Specifications
- Primary antibody reactivity:Human
- Description:Human JAM-A ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-502
Specifications
About this item
Human JAM-A AccuSignal ELISA Kit
- Natural and recombinant human JAM-A.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S28-V238
Junctional adhesion molecule A(JAM-A) is a protein that in humans is encoded by the F11R gene. It is mapped to 1q23.3. This gene is an immunoglobulin-like molecule that colocalizes with tight junctions in endothelium and epithelium and is also found on blood leukocytes and platelets. JAM-A plays an important role in the regulation of tight junction assembly in epithelia. In addition, it can act as a receptor for reovirus, a ligand for the integrin LFA1, involved in leukocyte transmigration, and a platelet receptor. JAM-A has a nonredundant role in controlling DC motility, trafficking to lymph nodes, and activation of specific immunity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419.pg/ml, 187.5pg/ml, 93.8pg/ml, 46.9pg/ml human JAM-A standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants or serum to each empty well. We recommend that each human JAM-A standard solution and each sample is measured in duplicate.