- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse Osteoactivin - GPNMB ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-188
Mouse Osteoactivin - GPNMB AccuSignal ELISA Kit
- Natural and recombinant mouse Osteoactivin.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: K23-N502
Transmembrane glycoprotein NMB is a protein that is encoded by the GPNMB gene. In osteoblast progenitor cells, GPNMB works as a positive regulator of osteoblast differentiation during later stages of matrix maturation and mineralization that is mediated at least in part by BMP-2 in a SMAD1 dependent manner to promote osteoblast differentiation. GPNMB can enhance the repairing process in bone fracture, demonstrated by its high expression during chondrogenesis(soft callus) and osteogenesis(hard callus) compared to the intact femurs that is why Osteoactivin(OA) could be a novel therapeutic agent used to treat generalized osteoporosis or localized osteopenia during fracture repair by stimulating bone growth and regeneration. Similarly, GPNMB expression increases during osteoclast differentiation and it is functionally implicated in this process, possibly by promoting the fusion of osteoclast progenitor cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml mouse Osteoactivin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Osteoactivin standard solution and each sample is measured in duplicate.