Order Entry
Canada
ContactUsLinkComponent
Mouse CD32 - FCGR2b - c ELISA Kit, Rockland Immunochemicals
Mouse CD32 - FCGR2b - c ELISA Kit, Rockland Immunochemicals
Catalog # 76236-184
CAS Number:  
Mouse CD32 - FCGR2b - c ELISA Kit, Rockland Immunochemicals
Catalog # 76236-184
Supplier Number:  KOA0786
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse CD32 - FCGR2b - c ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-184

Specifications

About this item

Mouse CD32 - FCGR2b - c AccuSignal ELISA Kit

  • Natural and recombinant mouse CD32.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T40-R217

CD32 is a surface receptor protein and part of a large population of B cell co-receptors, which act to modulate signaling. It has a low-affinity for IgG antibodies and down-regulates antibody production in the presence of IgG. This feedback loops essentially lowers the production of IgG produced by B cells when there is a surplus in the body.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse CD32 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD32 standard solution and each sample be measured in duplicate.