Order Entry
Canada
ContactUsLinkComponent
Human CD32 - FCGR2b - c ELISA Kit, Rockland Immunochemicals
Human CD32 - FCGR2b - c ELISA Kit, Rockland Immunochemicals
Catalog # 76236-182
CAS Number:  
Human CD32 - FCGR2b - c ELISA Kit, Rockland Immunochemicals
Catalog # 76236-182
Supplier Number:  KOA0785
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CD32 - FCGR2b - c ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-182

Specifications

About this item

Human CD32 - FCGR2b - c AccuSignal ELISA Kit

  • Natural and recombinant human CD32.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A46-P217

CD32 is a surface receptor protein and part of a large population of B cell co-receptors, which act to modulate signaling. It has a low-affinity for IgG antibodies and down-regulates antibody production in the presence of IgG. This feedback loops essentially lowers the production of IgG produced by B cells when there is a surplus in the body.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419.pg/ml, 187.5pg/ml, 93.8pg/ml, 46.9pg/ml human CD32 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human CD32 standard solution and each sample is measured in duplicate.