Order Entry
Canada
ContactUsLinkComponent
Rat TIMP-2 ELISA Kit, Rockland Immunochemicals
Rat TIMP-2 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-164
CAS Number:  
Rat TIMP-2 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-164
Supplier Number:  KOA0776
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rat
  • Target protein:
    TIMP-2
  • Description:
    Rat TIMP-2 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-164

Specifications

About this item

Rat TIMP-2 AccuSignal ELISA Kit

  • Natural and recombinant rat TIMP-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: C27-P220

TIMP-2 gen is encoded by 5 exons spanning 83 kb of genomic DNA. TIMP-2 is 83 kilobase pairs (kb) long with exon-intron splicing sites located in preserved positions among the three members of the TIMP family. The gene for tissue inhibitor of metalloproteinases-2 is localized on human chromosome arm 17q25. TIMP-2 abrogates angiogenic factor-induced endothelial cell proliferation in vitro and angiogenesis in vivo independent of MMP inhibition. The standard product used in this kit is recombinant human TIMP-2 with the molecular mass of 22Kda and 194 amino acid.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml rat TIMP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each rat TIMP-2 standard solution and each sample is measured in duplicate.