Specifications
- Target protein:IGFBP-5
- Description:Rat IGFBP5 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-484
Specifications
About this item
Rat IGFBP5 AccuSignal ELISA Kit
- Natural and recombinant rat IGFBP5.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: L20-E271
Insulin-like growth factor-binding protein 5 is a protein that in humans is encoded by the IGFBP5 gene. The IGFBP5 gene was mapped to chromosome 2q33-q34. The expression of IGFBP5 by stable transfection and adenovirus-mediated infection was inhibitory to growth in 2 human breast cancer cell lines. Stable expression of IGFBP5 in the breast cancer cell lines also inhibited the formation and growth of tumors following injection in athymic mice. IGFBP5 is believed a growth inhibitor and proapoptotic agent in breast cancer cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat IGFBP5 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat IGFBP5 standard solution and each sample be measured in duplicate.