ContactUsLinkComponent
Human REG-4 ELISA Kit, Rockland Immunochemicals
Human REG-4 ELISA Kit, Rockland Immunochemicals
  76237-470
 :  
Human REG-4 ELISA Kit, Rockland Immunochemicals
  76237-470
 :  KOA0765
 :  
  • Primary antibody reactivity:
    Human
  • Description:
    Human REG-4 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-470
Human REG-4 AccuSignal ELISA Kit
  • Natural and recombinant human REG-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: D23-P158
Regenerating islet-derived protein 4 is a protein that in humans is encoded by the REG4 gene. It is mapped to 1p12. REG-4 gene contains 6 exons and that the exon structure is preserved among members of the REG gene family. This gene is able to maintain carbohydrate recognition activity in an acidic environment. REG-4 expression was significantly upregulated by inflammation and tissue injury associated with active Crohn disease and ulcerative colitis, and most of the colorectal tumors overexpressing REG-4 are mucinous tumors or neuroendocrine tumors.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human REG-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human REG-4 standard solution and each sample be measured in duplicate.