Order Entry
Canada
ContactUsLinkComponent
Human Galectin-4 ELISA Kit, Rockland Immunochemicals
Human Galectin-4 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-966
CAS Number:  
Human Galectin-4 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-966
Supplier Number:  KOA0751
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Galectin-4 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-966

Specifications

About this item

Human Galectin-4 AccuSignal ELISA Kit

  • Natural and recombinant human Galectin-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A2-I323

Galectin-4, also known as LGALS4 or GAL4, is a protein that in humans is encoded by the LGALS4 gene. It is mapped to 19q13.2. Galectins are a family of animal lectins defined by an affinity for beta-galactoside-containing saccharides and by shared sequence elements. Galectin-4 is expressed as a 38-kD protein in the human colon adenocarcinoma T84 cell line. Galectin-4 is an S-type lectin that is strongly underexpressed in colorectal cancer, and this gene plays a role in cell adhesion. It has been found that decreased Galectin-4 expression occurs early in colorectal carcinogenesis.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Galectin-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Galectin-4 standard solution and each sample be measured in duplicate.