Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse GAS6 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-870
Specifications
About this item
Mouse GAS6 AccuSignal ELISA Kit
- Natural and recombinant mouse GAS6.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: D115-P673
GAS6(Growth arrest-specific 6), also known as AXLLG or AXSF, is a human gene coding for the Gas6 protein. It is mapped to 13q34. GAS6 was originally found as a gene upregulated by growth arrested fibroblasts. This gene product is a gamma-carboxyglutamic acid(Gla)-containing protein which thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. What's more, the product of the GAS6 gene, a ligand for the AXL, MER, and TYRO3 tyrosine kinase receptors, is a vitamin K-dependent protein, structurally related to anticoagulant protein S but lacking its anticoagulant activity. This protein also shows great importance in the cardiovascular system.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse GAS6 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse GAS6 standard solution and each sample be measured in duplicate.