Order Entry
Canada
ContactUsLinkComponent
Mouse MSP - MST1 ELISA Kit, Rockland Immunochemicals
Mouse MSP - MST1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-856
CAS Number:  
Mouse MSP - MST1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-856
Supplier Number:  KOA0688
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse MSP - MST1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-856

Specifications

About this item

Mouse MSP - MST1 AccuSignal ELISA Kit

  • Natural and recombinant mouse MSP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Q19-E716

Macrophage Stimulating 1(SMT1), also known as Hepatocyte growth factor-like protein(HGFL) and MSP, is a protein that in humans is encoded by the MST1 gene. The HGFL gene was identified at the DNF15S2 locus on human chromosome 3(3p21). The RON tyrosine kinase), the receptor for MSP, is expressed on the ciliated epithelia of the mucociliary transport apparatus of the lung. Furthermore, MSP stimulated ciliary motility in these cells by activating RON.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse MSP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse MSP standard solution and each sample be measured in duplicate.