Specifications
- Primary antibody reactivity:Human
- Description:Human KLK13 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-970
Specifications
About this item
Human KLK13 AccuSignal ELISA Kit
- Natural and recombinant human KLK13.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: G17-Q277
Kallikrein-13 is a protein that in humans is encoded by the KLK13 gene. It belongs to the kallikrein subgroup of serine proteases, which have diverse physiologic functions in many tissues. By genomic sequence analysis, KLK13 gene is mapped in a 300-kb region on chromosome 19q13.3-q13.4. It has been shown that recombinant hK13 produced in yeast can cleave synthetic peptides after the arginine residue and some extracellular matrix components. However, its exact physiological substrates and functions remain obscure. Despite the lack of knowledge on the physiological function of hK13, several studies have demonstrated that hK13 is implicated with cancer of the breast and ovary and it can serve as a favorable prognostic biomarker for these malignancies.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human KLK13 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, tissue homogenates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human KLK13 standard solution and each sample be measured in duplicate.