Specifications
- Primary antibody reactivity:Mouse
- Target protein:IL-15RA
- Description:Mouse IL15RA ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-822
Specifications
About this item
Mouse IL15RA AccuSignal ELISA Kit
- Natural and recombinant mouse IL15RA.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: G33-K205
Interleukin 15 receptor, alpha subunit is a subunit of the interleukin 15 receptor which is encoded by the gene IL15RA in humans. The human IL15RA gene was mapped to chromosome 10p15-p14 by FISH and the mouse Il15ra gene was mapped to chromosome 2. The cytoplasmic domain of IL15R-alpha, like that of IL2R-alpha, was dispensable for mitogenic signaling, suggesting that the primary role of the alpha chains is to confer high-affinity binding. At high concentrations, IL15, like IL2, was able to signal through a complex of IL2R-beta and -gamma in the absence of the alpha subunit. The soluble receptor displayed high affinity for IL15 and inhibited both binding of IL15 to the membrane receptor and IL15-induced cell proliferation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse IL15RA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL15RA standard solution and each sample be measured in duplicate.