Order Entry
Canada
ContactUsLinkComponent
 
Rat TGF-beta 3 ELISA Kit, Rockland Immunochemicals
  76235-782
 :  
Rat TGF-beta 3 ELISA Kit, Rockland Immunochemicals
  76235-782
 :  KOA0630
 :  

 

  • Assay Type:
    Sandwich
  • Description:
    Rat TGF-beta 3 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-782

 

 

Rat TGF-beta 3 AccuSignal ELISA Kit

  • Natural and recombinant rat TGF-beta 3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: A301-S413

Transforming growth factor beta 3(TGF-beta 3) is a type of protein, known as a cytokine, which is involved in cell differentiation, embryogenesis and development. It belongs to a large family of cytokines called the Transforming growth factor beta superfamily. TGF-beta 3 is believed to regulate molecules involved in cellular adhesion and extracellular matrix(ECM) formation during the process of palate development. Without TGF-beta 3, mammals develop a deformity known as acleft palate. This is caused by failure of epithelial cells in both sides of the developing palate to fuse. TGF-beta 3 also plays an essential role in controlling the development of lungs in mammals, by also regulating cell adhesion and ECM formation in this tissue, and controls wound healing by regulating the movements of epidermal and dermal cells in injured skin. TGF-beta 3 activated Lef1 in the absence of beta-catenin(CTNNB1) via nuclear phospho-Smad2 and Smad4.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml rat TGF-beta 3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of activated rat cell culture supernates, serum, plasma(EDTA) or urine to each empty well. It is recommended that each rat TGF-beta 3 standard solution and each sample be measured in duplicate.