- Primary antibody reactivity:Human
- Description:Human Seprase - FAP ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-776
Human Seprase - FAP AccuSignal ELISA Kit
- Natural and recombinant human Seprase.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: L26-D760
FAP(Fibroblast Activation Protein, Alpha) also known as FAPA or SEPRASE, is an inducible cell surface glycoprotein that was originally identified in cultured fibroblasts using monoclonal antibody F19. The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. The FAP gene is mapped on 2q24.2. FAP is most closely related to DPPIV and they share about 50% of their amino acids. FAP is catalytically active as a 170kD dimer and has dipeptidase and gelatinase activity. Its gelatinase activity requires a glycine in P2 position.FAP-alpha shows 48% amino acid identity with dipeptidyl peptidase IV and 30% identity with DPP4-related protein. Northern blot analysis detected a 2.8-kb FAP-alpha mRNA in fibroblasts. Depletion of FAP-expressing cells, which made up only 2% of all tumor cells in established Lewis lung carcinomas, caused rapid hypoxic necrosis of both cancer and stromal cells in immunogenic tumors by a process involving interferon-gamma and tumor necrosis factor-alpha.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human Seprase standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Seprase standard solution and each sample be measured in duplicate.