Specifications
- Primary antibody reactivity:Mouse
- Target protein:MMP-12
- Description:Mouse MMP-12 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-766
Specifications
About this item
Mouse MMP-12 AccuSignal ELISA Kit
- Natural and recombinant mouse MMP-12.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: A18-C462
Matrix metalloproteinase-12(MMP12), also known as MME or ME, is an enzyme that in humans is encoded by the MMP12 gene. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.2. It is thought that the protein encoded by this gene is cleaved at both ends to yield the active enzyme, but this processing has not been fully described. The enzyme degrades soluble and insoluble elastin. It may play a role in aneurysm formation and studies in mice suggest a role in the development of emphysema. This gene may involved in tissue injury and remodeling.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse MMP-12 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each mouse MMP-12 standard solution and each sample be measured in duplicate.